MSD READ BUFFER T (4X
Catalog # MESOR92TC-1
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
STREPTAVIDON SULFO-TAG LABELED
Supplier: MESO SCALE DISCOVERY
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Capillary pistons for Microman® pipettes
Supplier: GILSON
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Magnetic stirring bars, micro
Supplier: VWR Collection
Kits for Precellys®
Supplier: Bertin Instruments
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
EasySep™ Human CD8+ T Cell Isolation Kits
Supplier: STEMCELL Technologies