VWR® Basic, Aprons, PE
White, 800×1400 mm
113-9094
Disposable aprons made of PE film.
- Free from natural rubber latex
- Available in two different sizes
Thickness: 20 µm
: 50 pieces per bag, 10 bags per case.
- Colour:White
- W×L:800×1400 mm
- Thickness:20 µm
- Cat. No.:113-9094
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



