Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Basic, Shoe Covers

VWR® Basic, Shoe Covers

40 µm, Green, U*
Catalog # 113-9086
Supplier:   VWR Collection
VWR® Basic, Shoe Covers
Catalog # 113-9086
Supplier:  VWR Collection
VWR® Basic, Shoe Covers
VWR® Basic, Shoe Covers

40 µm, Green, U*

Price
68,60 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Disposable shoe covers made of CPE film.

  • Free from natural rubber latex
  • Available in different colours

L×W: 410×170 mm

Thickness: 25, 40 or 70 µm

* One size fits all

Packaging: 10 pieces per bundle, 10 bundles per bag, 10 bags per case.

Specifications
  • Shoe size:
    U*
  • Colour:
    Green
  • Description:
    40 µm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.