ContactUsLinkComponent
Blasticidin S hydrochloride, 10 mg/ml in HEPES buffer, sterile-filtered 1 * 1 mL
Catalog # J67216.XF
Blasticidin S hydrochloride, 10 mg/ml in HEPES buffer, sterile-filtered 1 * 1 mL
Catalog # J67216.XF
Frequently Bought Together