Buffer LBP (125 mL) Lysis Buffer LBP bottle of 125 mL no dangerous goods according transport regulations 1 * 125 mL
MANA740906.125
Buffer LBP (125 mL) Lysis Buffer LBP bottle of 125 mL no dangerous goods according transport regulations 1 * 125 mL
MANA740906.125
Frequently Bought Together
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Single use syringes, 2-piece, Injekt® Solo
: B. Braun
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
qScript™ XLT One-Step RT-qPCR ToughMix®
: Quantabio
VWR® Standard Line Transfer Pipettes
: VWR Collection
Decontamination solution, DNA AWAY®
: Thermo Fisher Scientific
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
: EPPENDORF
Single-use syringes, 3-piece, HENKE-JECT®
: HENKE SASS WOLF