NucleoVac 24 Vacuum Manifold Vacuum manifold for processing 1–24 NucleoSpin or NucleoSnap columns - manifold, NucleoVac Mini Adapter , Luer plugs, tubing connection, closing plug 1 * 1 SET
Catalog # MANA740299
Supplier: MACHEREY-NAGEL
Frequently Bought Together