Trolleys, serving
Serving trolley, heavy duty, with reinforced chassis, SSW, 895×595×1015 mm, 28,13 kg, 3
HUPF0112322
Transport trolley complete with 25 mm deep shelves. Shelves complete with bevelled edges for user comfort. With four swivel castors, two of them with brake.
- Corrosion resistant
- Disc bumpers are made of elastic and shock resistant plastic material, it protect trolley from damage
- Maintenance free
High-quality stainless steel construction.
- Description:Serving trolley, heavy duty, with reinforced chassis, SSW
- W×D×H:895×595×1015 mm
- Shelves:3
- Weight:28,13 kg
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.



