ContactUsLinkComponent
Seal heat sealable PierceASeal Foil Super 500m x 115mm pierceable foil seal with moderate solvent resistance peelable and piercable easy sealing surface identification non-sterile 1 * 1 Roll
Catalog # ISTSIST-115-115LR
Supplier:  iST Scientific Ltd
Seal heat sealable PierceASeal Foil Super 500m x 115mm pierceable foil seal with moderate solvent resistance peelable and piercable easy sealing surface identification non-sterile 1 * 1 Roll
Catalog # ISTSIST-115-115LR
Supplier:  iST Scientific Ltd
Frequently Bought Together