VWR® Basic, Overalls, with Hood
White, XL
Catalog # 113-9029
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable overalls, with hood, are made from non woven PE-laminated, PP fabric.
- Hood
- Zip closure (one direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Verpakking: 1 piece per bag, 50 bags per case.
Documents
CoC Overalls, with hood, VWR® Basic CoC_VWRI113-9027_26320182 CoC_VWRI113_Overalls, with Hood, VWR® Basic VWRI113-9035_04520252.pdf
Food contact certificate F
Aanvullende productdetails
TDS VWR® Coverall Laminated PP Hood with Zip TDS VWR® Coverall Laminated PP Hoods with Zip
Technische fiche
VWR® Non Regulated Disposables for Production
Product documentatie
Specifications
- Clothing Size:XL
- Colour:White
- Seq:04
- Cat. No.:113-9029
- Garment application:General Purpose|Laboratory
- Weight:40 g/m²
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.