Orders LinkContactUsLinkComponent
VWR® Basic, Overalls, with Hood
VWR® Basic, Overalls, with Hood
Blue, XL
Catalogus # 113-9023
Leverancier:   VWR Collection
VWR® Basic, Overalls, with Hood
Catalogus # 113-9023
Leverancier:  VWR Collection
VWR® Basic, Overalls, with Hood
VWR® Basic, Overalls, with Hood
Blue, XL
Price
289,00 €
Log in voor beschikbaarheid
1 * 50 ST
Product Details & Documents
These disposable overalls, with hood, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
  • Hood
  • Zip closure (one direction)
  • Elasticated wrists
  • Free from natural rubber latex
Weight: 40 g/m²
Verpakking: 1 piece per bag, 50 bags per case.
Specificaties
  • Clothing Size:
    XL
  • Colour:
    Blue
  • Seq:
    10
  • Cat. No.:
    113-9023
  • Garment application:
    General Purpose|Laboratory
  • Weight:
    40 g/m²
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.