Orders LinkContactUsLinkComponent
VWR® Basic, Overalls, with Hood
VWR® Basic, Overalls, with Hood
White, S
Catalog # 113-9014
Supplier:   VWR Collection
VWR® Basic, Overalls, with Hood
Catalog # 113-9014
Supplier:  VWR Collection
VWR® Basic, Overalls, with Hood
VWR® Basic, Overalls, with Hood
White, S
Price
289,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
These disposable overalls, with hood, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
  • Hood
  • Zip closure (one direction)
  • Elasticated wrists
  • Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Specifications
  • Clothing Size:
    S
  • Colour:
    White
  • Seq:
    01
  • Cat. No.:
    113-9014
  • Garment application:
    General Purpose|Laboratory
  • Weight:
    40 g/m²