VWR® Basic, Overalls, Dual-Zip Closure
White, XXL
Catalog # 113-9013
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Collar
- Zip closure (dual-direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
Food contact certificate F
Additional Product Details
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Clothing Size:XXL
- Colour:White
- Seq:04
- Cat. No.:113-9013
- Garment application:General Purpose|Laboratory|Industrial
- Closure type:Zip closure
- Weight:40 g/m²



