VWR® Basic, Overalls, Dual-Zip Closure
White, XL
Catalog # 113-9012
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Collar
- Zip closure (dual-direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
Food contact certificate F
Additional Product Details
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Clothing Size:XL
- Colour:White
- Seq:03
- Cat. No.:113-9012
- Garment application:General Purpose|Laboratory|Industrial
- Closure type:Zip closure
- Weight:40 g/m²
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



