Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Basic, Overalls

VWR® Basic, Overalls

Blue, 3XL
Catalog # 113-9009
Supplier:   VWR Collection
VWR® Basic, Overalls
Catalog # 113-9009
Supplier:  VWR Collection
VWR® Basic, Overalls
VWR® Basic, Overalls

Blue, 3XL

Price
125,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.

  • Collar
  • Zip closure (one direction)
  • Elasticated wrists
  • Free from natural rubber latex

Weight: 40 g/m²

Packaging: 1 piece per bag, 50 bags per case.

Specifications
  • Clothing Size:
    3XL
  • Colour:
    Blue
  • Seq:
    08
  • Cat. No.:
    113-9009
  • Garment application:
    General Purpose|Laboratory
  • Weight:
    40 g/m²