Orders LinkContactUsLinkComponent
VWR® Basic, Overalls
VWR® Basic, Overalls
Blue, XXL
N° de catalogue 113-9008
Fournisseur:   VWR Collection
VWR® Basic, Overalls
N° de catalogue 113-9008
Fournisseur:  VWR Collection
VWR® Basic, Overalls
VWR® Basic, Overalls
Blue, XXL
Prix
125,00 €
Connexion Pour la disponibilité
1 * 50 ST
Product Details & Documents
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
  • Collar
  • Zip closure (one direction)
  • Elasticated wrists
  • Free from natural rubber latex
Weight: 40 g/m²
Emballage: 1 piece per bag, 50 bags per case.
Spécifications
  • Clothing Size:
    XXL
  • Colour:
    Blue
  • Seq:
    07
  • Cat. No.:
    113-9008
  • Garment application:
    General Purpose|Laboratory
  • Weight:
    40 g/m²