Orders LinkContactUsLinkComponent
VWR® Basic, Overalls
VWR® Basic, Overalls
White, XL
  113-9003
VWR® Basic, Overalls
  113-9003
VWR® Basic, Overalls
VWR® Basic, Overalls
White, XL
125,00 €
1 * 50 ST
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
  • Collar
  • Zip closure (one direction)
  • Elasticated wrists
  • Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
  • Clothing Size:
    XL
  • Colour:
    White
  • Seq:
    02
  • Cat. No.:
    113-9003
  • Garment application:
    General Purpose|Laboratory
  • Weight:
    40 g/m²