VWR® Basic, Frocks, Zip Closure
White, XXL, 112 cm
113-9001
These disposable frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
- Zip closure
- Elastic wrists
- Raglan sleeves
- Free from natural rubber latex
Weight: 40 g/m²
: 1 piece per bag, 50 bags per case.
- Clothing Size:XXL
- Colour:White
- Length:112 cm
- Seq:02
- Cat. No.:113-9001
- Garment application:General Purpose|Laboratory
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Zip closure
- Weight:40 g/m²



