Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Basic, Frocks, Zip Closure

VWR® Basic, Frocks, Zip Closure

White, XL, 108 cm
Catalog # 113-9000
Supplier:   VWR Collection
VWR® Basic, Frocks, Zip Closure
Catalog # 113-9000
Supplier:  VWR Collection
VWR® Basic, Frocks, Zip Closure
VWR® Basic, Frocks, Zip Closure

White, XL, 108 cm

Price
95,70 €
Log in for availability
1 * 50 ST
Product Details & Documents

These disposable frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.

  • Zip closure
  • Elastic wrists
  • Raglan sleeves
  • Free from natural rubber latex

Weight: 40 g/m²

Packaging: 1 piece per bag, 50 bags per case.

Specifications
  • Clothing Size:
    XL
  • Colour:
    White
  • Length:
    108 cm
  • Seq:
    01
  • Cat. No.:
    113-9000
  • Garment application:
    General Purpose|Laboratory
  • Gender:
    Unisex
  • Pockets:
    Without pockets
  • Closure type:
    Zip closure
  • Weight:
    40 g/m²