VWR® Basic, Frocks
White, XL, 108 cm
Catalog # 113-8996
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
- Press stud fastening
- Elastic wrists
- Raglan sleeves
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
Food contact certificate F
Additional Product Details
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Clothing Size:XL
- Colour:White
- Length:108 cm
- Seq:02
- Cat. No.:113-8996
- Garment application:General Purpose|Laboratory
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Press studs / snaps
- Weight:40 g/m²
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



