Orders LinkContactUsLinkComponent
VWR® Basic, Beard Covers
VWR® Basic, Beard Covers
White
Catalog # 113-8902
Supplier:   VWR Collection
VWR® Basic, Beard Covers
Catalog # 113-8902
Supplier:  VWR Collection
VWR® Basic, Beard Covers
VWR® Basic, Beard Covers
White
Price
104,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
This disposable beard cover is made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in white and blue
Weight: 10 g/m²
Packaging: 100 pieces/bag, 10 bags/case.
Specifications
  • Colour:
    White
  • Cat. No.:
    113-8902
  • Weight:
    10 g/m²