Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Basic Mob Caps, Single Elastic

VWR® Basic Mob Caps, Single Elastic

Green, 580 mm
Catalog # 113-8858
Supplier:   VWR Collection
VWR® Basic Mob Caps, Single Elastic
Catalog # 113-8858
Supplier:  VWR Collection
VWR® Basic Mob Caps, Single Elastic
VWR® Basic Mob Caps, Single Elastic

Green, 580 mm

Price
49,70 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

These disposable pleated caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.

  • Free from natural rubber latex
  • Single elastic
  • Available in a wide range of colours

Weight: 10 g/m²

Packaging: 100 pieces per bag, 10 bags per case.

Specifications
  • Ø:
    580 mm
  • Colour:
    Green