Orders LinkContactUsLinkComponent
VWR®, IOS W-Plan Objectives
VWR®, IOS W-Plan Objectives
IOS W-Plan objective (oil) with iris for Darkfield
Catalog # 630-2954
Supplier:   VWR Collection
VWR®, IOS W-Plan Objectives
Catalog # 630-2954
Supplier:  VWR Collection
VWR®, IOS W-Plan Objectives
IOS W-Plan objective (oil) with iris for Darkfield
Price
612,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These high contrast objectives are ideal for routine observations in laboratories.
  • Ensure field flatness up to F.N. 22
  • Based on infinity-corrected optical system
  • 100× is also available with iris for darkfield
Documents
Specifications
  • Magnification:
    100×/1,25 (i)
  • Description:
    IOS W-Plan objective (oil) with iris for Darkfield
  • Seq:
    157
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.