Orders LinkContactUsLinkComponent
VWR®, IOS W-Plan Objectives
VWR®, IOS W-Plan Objectives
IOS W-Plan objective
Catalog # 630-2948
Supplier:   VWR Collection
VWR®, IOS W-Plan Objectives
Catalog # 630-2948
Supplier:  VWR Collection
VWR®, IOS W-Plan Objectives
IOS W-Plan objective
Price
110,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These high contrast objectives are ideal for routine observations in laboratories.
  • Ensure field flatness up to F.N. 22
  • Based on infinity-corrected optical system
  • 100× is also available with iris for darkfield
Documents
Specifications
  • Magnification:
    2×/0,05
  • Description:
    IOS W-Plan objective
  • Seq:
    150