ContactUsLinkComponent
Wearing Disc P-MRK, PTFE-PDR, suitable for Magnetic Stirrer Heads P-MRK with Ground Joint NS 29/32 for shaft O. D. 8 mm, #C520-24 as well as Magnetic Stirrer Heads P-MRK with Ground Joint NS 29/32 or NS 45/40 for shaft O. D. 10 mm, #C520-28 and C520- 1 * 3 items
Catalog # BOHLC931-01
Supplier:  Bohlender
undefined
Wearing Disc P-MRK, PTFE-PDR, suitable for Magnetic Stirrer Heads P-MRK with Ground Joint NS 29/32 for shaft O. D. 8 mm, #C520-24 as well as Magnetic Stirrer Heads P-MRK with Ground Joint NS 29/32 or NS 45/40 for shaft O. D. 10 mm, #C520-28 and C520- 1 * 3 items
Catalog # BOHLC931-01
Supplier:  Bohlender
Frequently Bought Together