ContactUsLinkComponent
Tubing Precision Clean II, made of clear amine-free Low Density PolyEthylene, suitable for use in cleanrooms, manufactured and packed in an ISO 4 cleanroom, thickness 2mil (50 micron) per layer, width/opening 20 (508mm), length 1000feet (305m), 1 ro 1 * 1 Roll
Catalog # FISH1200200
Supplier:  FISHER CONTAINER
Tubing Precision Clean II, made of clear amine-free Low Density PolyEthylene, suitable for use in cleanrooms, manufactured and packed in an ISO 4 cleanroom, thickness 2mil (50 micron) per layer, width/opening 20 (508mm), length 1000feet (305m), 1 ro 1 * 1 Roll
Catalog # FISH1200200
Supplier:  FISHER CONTAINER
Frequently Bought Together