ContactUsLinkComponent
Bag Precision Clean II, made of clear amine-free Low Density PolyEthylene, suitable for use in cleanrooms, manufactured and packed in an ISO 4 cleanroom, thickness 4mil (100 micron) per layer, width/opening 10 (254mm), length 12 (305mm), 1000 piece 1 * 1.000 items
Catalog # FISH11030
Supplier:  FISHER CONTAINER
Bag Precision Clean II, made of clear amine-free Low Density PolyEthylene, suitable for use in cleanrooms, manufactured and packed in an ISO 4 cleanroom, thickness 4mil (100 micron) per layer, width/opening 10 (254mm), length 12 (305mm), 1000 piece 1 * 1.000 items
Catalog # FISH11030
Supplier:  FISHER CONTAINER
Frequently Bought Together