ContactUsLinkComponent
Bag Precision Clean II, made of clear amine-free Low Density PolyEthylene, suitable for use in cleanrooms, manufactured and packed in an ISO 4 cleanroom, thickness 4mil (100 micron) per layer, width/opening 8 (203mm), length 10 (254mm), 1000 pieces 1 * 1.000 items
Catalog # FISH11029
Supplier:  FISHER CONTAINER
Bag Precision Clean II, made of clear amine-free Low Density PolyEthylene, suitable for use in cleanrooms, manufactured and packed in an ISO 4 cleanroom, thickness 4mil (100 micron) per layer, width/opening 8 (203mm), length 10 (254mm), 1000 pieces 1 * 1.000 items
Catalog # FISH11029
Supplier:  FISHER CONTAINER
Frequently Bought Together