User maintenance kit for Precellys Evolution is made for preventive maintenance and contains:#2 x vacuum joint#1 x containment seal# 1 x antirotation tubing kit 1 * 1 items
Catalog # BERTS000808-PEVO0A
Supplier: Bertin Instruments
User maintenance kit for Precellys Evolution is made for preventive maintenance and contains:#2 x vacuum joint#1 x containment seal# 1 x antirotation tubing kit 1 * 1 items
Catalog # BERTS000808-PEVO0A
Supplier: Bertin Instruments
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Clean-Lab, Q Path® for paraffin removal
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Safety tube holders, VACUETTE® QUICKSHIELD
Supplier: Greiner Bio-One