ContactUsLinkComponent
This reference includes all the spare parts on Precellys 24-Dual needed to be changed on a yearly basis as discribed in the user man ual. 1 x white blocking plate, 2 x vaccuum joint, 1 x containment seal, 1 x antirotation tubing kit. 1 * 1 KIT
Catalog # 432-0247
Supplier:  Bertin Instruments
This reference includes all the spare parts on Precellys 24-Dual needed to be changed on a yearly basis as discribed in the user man ual. 1 x white blocking plate, 2 x vaccuum joint, 1 x containment seal, 1 x antirotation tubing kit. 1 * 1 KIT
Catalog # 432-0247
Supplier:  Bertin Instruments
Frequently Bought Together