2-Phenylethanol USP
15,9 kg
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
- CAS Number:60-12-8
- Formula:C₆H₅CH₂CH₂OH
- MDL Number:MFCD00002886
- Molecular Weight:122.17 g/mol
- Merck Index:12,07370
- Boiling Pt:432 °C (1013 hPa)
- Melting Pt:103…105 °C
- Density:1.02 g/cm³ (20 °C)
- Flash Pt:102 °C
- Storage Temperature:Ambient
- EINECS:200-456-2
- UN:2810
- ADR:6.1,III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:15,9 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



