High Stackable Ledges, 100 mm
Shelf ledge, Metroseal green with Microban®, 530 mm width
METOL21N-4K3
Actual ledge length is approximately 25 mm shorter than nominal shelf length/width.
- Description:Shelf ledge, Metroseal green with Microban®, 530 mm width
- Seq:02



