Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Xylan (from corn core)

100 g
  TCIAX0078-100G
 :   TCI
Xylan (from corn core)
  TCIAX0078-100G
 :  TCI
 :  9014-63-5
Xylan (from corn core)

100 g

 :  9014-63-5
 
128,00 €
   
1 * 100 g
 
  •  :9014-63-5
  •  :MFCD00082148
  •  :232-760-6
 

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Xylose:
    min. 75.0 % (after hydrolysis)
  • Drying loss:
    max. 15.0 %

 

  • Pk:
    100 g