Order Entry
Belgium
Orders LinkContactUsLinkComponent
Nile red

Nile red

5 g
Catalog # TCIAN0659-5G
Supplier:   TCI
Nile red
Catalog # TCIAN0659-5G
Supplier:  TCI
CAS Number:  7385-67-3
Nile red
Nile red

5 g

CAS Number:  7385-67-3
Price
469,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
  • CAS Number:7385-67-3
  • Formula:C₂₀H₁₈N₂O₂
  • MDL Number:MFCD00011639
  • Molecular Weight:318.38 g/mol
  • EINECS:230-966-0
Specifications

Specification Test Results

  • Appearance:
    Yellow to Brown to Dark green powder to crystal
  • Lambda max.:
    550.0 to 554.0 nm (MeOH)
  • Absorbance (E1%1cm):
    min. 1,300 (MeOH, 550.0 to 554.0 nm)

Specifications

  • Pk:
    5 g

Specifications