ContactUsLinkComponent
Amarell-Special-Therm., -58+5:0,1°C, enclosed scale, cap. round, colourless, toluene filled, c.450x9,0-9,5mm, Richter-top finish, g overnment verified, null
Catalog # AMARN61618-TOL/CC
Supplier:  Amarell
Amarell-Special-Therm., -58+5:0,1°C, enclosed scale, cap. round, colourless, toluene filled, c.450x9,0-9,5mm, Richter-top finish, g overnment verified, null
Catalog # AMARN61618-TOL/CC
Supplier:  Amarell
Frequently Bought Together