Therm. similar Landsberger, +34+52:0,05°C, enclosed scale, capillary tube specially coated inside, with non-wetting blue special li quid, total immersion, c.300x9mm, Richter-top finish with button, null
Catalog # AMARN63804-CB-CC
Supplier: Amarell
Frequently Bought Together