ContactUsLinkComponent
Amarell-Special-Therm., +198+252:0,1°C, enclosed scale, cap. round, colourless, gallium filled, c.450x9,0-9,5mm, Richter-top finish, with ice point scale, suitable for government calibration 1 * 1 items
Catalog # AMARN61628-GA
Supplier:  Amarell
Amarell-Special-Therm., +198+252:0,1°C, enclosed scale, cap. round, colourless, gallium filled, c.450x9,0-9,5mm, Richter-top finish, with ice point scale, suitable for government calibration 1 * 1 items
Catalog # AMARN61628-GA
Supplier:  Amarell
Frequently Bought Together