ContactUsLinkComponent
Therm., similar Dr. Otte, similar DIN, 0+250:1°C, enclosed scale, cap. prismatic, colourless, blue special liquid, 370x7mm, 16lll gl ass, Richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARN59804-FL
Supplier:  Amarell
Therm., similar Dr. Otte, similar DIN, 0+250:1°C, enclosed scale, cap. prismatic, colourless, blue special liquid, 370x7mm, 16lll gl ass, Richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARN59804-FL
Supplier:  Amarell
Frequently Bought Together