ContactUsLinkComponent
Crystallization point therm., similar DIN 12785, +4,0+6,0:0,01°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, 250x6mm, null
Catalog # AMARM253590-CB
Supplier:  Amarell
Crystallization point therm., similar DIN 12785, +4,0+6,0:0,01°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, 250x6mm, null
Catalog # AMARM253590-CB
Supplier:  Amarell
Frequently Bought Together