ContactUsLinkComponent
Floating therm., 0+100:1C, cap. lens front, colourless, red special liquid, shot weighted, floating upright, ca. 250x17mm, total im mersion, rounded top finish 1 * 1 items
  AMART108510-FL
 :  Amarell
Floating therm., 0+100:1C, cap. lens front, colourless, red special liquid, shot weighted, floating upright, ca. 250x17mm, total im mersion, rounded top finish 1 * 1 items
  AMART108510-FL
 :  Amarell
Frequently Bought Together