ContactUsLinkComponent
Quartz-Glass therm., 0+800:5°C, upper part c. 300x13-15mm, lower part 1000x8-9mm, milk-glass scale, cap. colourless, gallium filled, partial immersion is by 5-10cm shorter than the stem, cork-plaster-top finish, with metal cap, suitable for governme 1 * 1 items
Catalog # AMARQ137120-GA
Supplier:  Amarell
Quartz-Glass therm., 0+800:5°C, upper part c. 300x13-15mm, lower part 1000x8-9mm, milk-glass scale, cap. colourless, gallium filled, partial immersion is by 5-10cm shorter than the stem, cork-plaster-top finish, with metal cap, suitable for governme 1 * 1 items
Catalog # AMARQ137120-GA
Supplier:  Amarell
Frequently Bought Together