ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 4d, solid stem, white backed, -2+400:1°C, length 430mm, imm. 100mm, gallium filled, durable pigm ent, suitable for government verification 1 * 1 items
Catalog # AMARA400020-GA
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 4d, solid stem, white backed, -2+400:1°C, length 430mm, imm. 100mm, gallium filled, durable pigm ent, suitable for government verification 1 * 1 items
Catalog # AMARA400020-GA
Supplier:  Amarell
Frequently Bought Together