ContactUsLinkComponent
Thermometer similar to IP 53 C, solid stem, white backed, 0+80:0,5°C, red special liquid, durable pigment, suitable for government v erification 1 * 1 items
Catalog # AMARA340780-FL
Supplier:  Amarell
Thermometer similar to IP 53 C, solid stem, white backed, 0+80:0,5°C, red special liquid, durable pigment, suitable for government v erification 1 * 1 items
Catalog # AMARA340780-FL
Supplier:  Amarell
Frequently Bought Together