ContactUsLinkComponent
Thermometer similar to IP 52 C, solid stem, white backed, 90+260:1°C, gallium filled, durable pigment, suitable for government verif ication 1 * 1 items
Catalog # AMARA340760-GA
Supplier:  Amarell
Thermometer similar to IP 52 C, solid stem, white backed, 90+260:1°C, gallium filled, durable pigment, suitable for government verif ication 1 * 1 items
Catalog # AMARA340760-GA
Supplier:  Amarell
Frequently Bought Together