ContactUsLinkComponent
Hydrometer, DIN 12791/similar to BS 718, L20, 0,9800-1,0000:0,0002g/cm , ref. temp. 20C, without therm., 430mm long, suitable for g overnment verification 1 * 1 items
Catalog # AMARH805038
Supplier:  Amarell
Hydrometer, DIN 12791/similar to BS 718, L20, 0,9800-1,0000:0,0002g/cm , ref. temp. 20C, without therm., 430mm long, suitable for g overnment verification 1 * 1 items
Catalog # AMARH805038
Supplier:  Amarell
Frequently Bought Together