ContactUsLinkComponent
Thermometer similar to IP 9 C, solid stem, white backed, 40+85:0,2C, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA340110-CB
Supplier:  Amarell
Thermometer similar to IP 9 C, solid stem, white backed, 40+85:0,2C, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA340110-CB
Supplier:  Amarell
Frequently Bought Together