ContactUsLinkComponent
Refill therm. for silo and pit therm., 0+100:1C, special red liquid filled, milk glass scale, cap. lens front, yellow backed, lower part 1000mm, partial immersion 1 * 1 items
Catalog # AMARK190088-FL
Supplier:  Amarell
Refill therm. for silo and pit therm., 0+100:1C, special red liquid filled, milk glass scale, cap. lens front, yellow backed, lower part 1000mm, partial immersion 1 * 1 items
Catalog # AMARK190088-FL
Supplier:  Amarell
Frequently Bought Together