ContactUsLinkComponent
Fadentherm., 0+100:1°C, solid stem, white backed, red special liquid, 300x6-7mm, bulb length 100mm, partial immersion, 16 B glass, w ith button at top, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARL44102-FL
Supplier:  Amarell
Fadentherm., 0+100:1°C, solid stem, white backed, red special liquid, 300x6-7mm, bulb length 100mm, partial immersion, 16 B glass, w ith button at top, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARL44102-FL
Supplier:  Amarell
Frequently Bought Together