ContactUsLinkComponent
Glycerine tester, 0-100:1%mas, with thermometer, 0+35:1C with red or blue special liquid, reference temperature 15C, 360mm long 1 * 1 items
Catalog # AMARH860720-FL
Supplier:  Amarell
Glycerine tester, 0-100:1%mas, with thermometer, 0+35:1C with red or blue special liquid, reference temperature 15C, 360mm long 1 * 1 items
Catalog # AMARH860720-FL
Supplier:  Amarell
Frequently Bought Together