Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hydrometer for milk of lime, 0-50g/l, without thermometer, reference temperature 15C, 260mm long 1 * 1 items
Catalog # AMARH860680
Supplier:
Amarell
Expand image
Hydrometer for milk of lime, 0-50g/l, without thermometer, reference temperature 15C, 260mm long 1 * 1 items
Catalog # AMARH860680
Supplier:
Amarell
Frequently Bought Together
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
VWR® Burettes, with PTFE Key Stopcock
Supplier: VWR Collection
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck