ContactUsLinkComponent
Stirring therm., enclosed scale, -10+250:1C, 300x7-8mm, cap. prismatic, colourless, red special liquid, total immersion, with ring at top 1 * 1 items
Catalog # AMARG11918-FL
Supplier:  Amarell
Stirring therm., enclosed scale, -10+250:1C, 300x7-8mm, cap. prismatic, colourless, red special liquid, total immersion, with ring at top 1 * 1 items
Catalog # AMARG11918-FL
Supplier:  Amarell
Frequently Bought Together